Applied Biosystems™ RNase Inhibitor DFS Item
No DNase or endonuclease activity
Supplier:  Applied Biosystems™ N8080119
Catalog No. N8080119
Explore available promotions
Includes: 100μL at 20 U/μL. Sufficient for 100 reverse transcription reactions, 20μL each. RNase Inhibitor in Storage Buffer: 20mM HEPES-KOH, pH 7.6; 50mM KCl; 8mM DTT; and 50% (v/v) glycerol.
Description
Description
RNase Inhibitor (ribonuclease inhibitor) is a 50 kDa recombinant enzyme used to inhibit RNase activity. It does not contain DNase or endonuclease activity.
  • Inhibits RNase activity—preventing degradation of RNA template
  • Lacks DNA endonuclease activity—for better product yield
PCR and Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
50 mM
Contents: 100 μL at 20 U/μL. Sufficient for 100 reverse transcription reactions, 20 μl each. RNase Inhibitor in Storage Buffer: 20 mM HEPES-KOH, pH 7.6; 50 mM KCl; 8 mM DTT; and 50% (v/v) glycerol. Store at -20°C.
2,000 units
Dry Ice
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.