missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ GTXpress™ Master Mix
Combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format.
Supplier: Applied Biosystems™ 4403311
Description
TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. The 100mL pack (2 x 50mL) includes reagents sufficient to perform 40,000 PCR reactions (5μL/reaction).
- Compatible with both Fast and Standard mode thermal cycling conditions
- Pre-mixed components stored at 2° to 8°C significantly reduce assay setup time
- Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
- Extended PCR cycling can be used for poorly amplified or sample-limited reactions The TaqMan™ GTXpress™ Master Mix is also sold as part of TaqMan™ Sample-to-SNP™ Kit, which provides real-time PCR-ready DNA in 5 min. from a wide variety of samples, ranging from blood to buccal swabs to plant tissues. For more information on the TaqMan™ Sample-to-SNP™ Kit, refer to the Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit Protocol.
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
| 2X | |
| Primer-probe | |
| High | |
| AmpliTaq DNA Polymerase | |
| SNP Genotyping (Real-Time PCR-Based) | |
| SNPs (Unknown or Numerous), SNPs (Known) | |
| GTXpress, TaqMan | |
| UP (Ultra Pure) | |
| Wet Ice | |
| 2 X | |
| 40,000 Reactions | |
| 100 mL |
| Contains 1 mL of TaqMan™ GTXpress™ Master Mix (2X) with AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye and ROX™ Dye. Sufficient for 400 reactions. Store at 2-8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| Single Vial | |
| qPCR | |
| Fast or Standard | |
| QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™ Standard Mode, StepOne™ Fast Mode | |
| ROX (Pre-mixed) | |
| Master Mix | |
| 1 mL | |
| Genotyping | |
| Built-In Hot Start | |
| DNA (Genomic), Sample-to-SNP™ Kit Lysates |
For Research Use Only. Not for use in diagnostic procedures.