missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ TaqMan™ Multiplex Master Mix DFS Item

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

Supplier:  Applied Biosystems™ 4461884

 View more versions of this product

Catalog No. 4461884


Explore available promotions

Description

Description

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
Specifications

Specifications

Contains 2 bottles (5 mL each) of 2X TaqMan™ Multiplex Master Mix sufficient for 1000 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

High
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Multiplex Master Mix
1000 Reactions
2X
1000 Reactions
10 mL
Primer-probe
Multiplex qPCR
Fast
TaqMan™
2 x 5 mL
Gene Expression, Virus Detection
2 X
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.