missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Apronal 98.0+%, TCI America™
$53.32 - $156.03
Chemical Identifiers
CAS | 528-92-7 |
---|---|
Molecular Formula | C9H16N2O2 |
Molecular Weight (g/mol) | 184.239 |
MDL Number | MFCD00210239 |
InChI Key | KSUUMAWCGDNLFK-UHFFFAOYSA-N |
Synonym | Apronalide, Allylisopropylacetylcarbamide, Allylisoprorylacetylurea, (2-Isopropyl-4-pentenoyl)urea, N-Carbamoyl-2-isopropyl-4-pentenamide |
PubChem CID | 10715 |
IUPAC Name | N-carbamoyl-2-propan-2-ylpent-4-enamide |
SMILES | CC(C)C(CC=C)C(=O)NC(=O)N |
Chemical Identifiers
528-92-7 | |
184.239 | |
KSUUMAWCGDNLFK-UHFFFAOYSA-N | |
10715 | |
CC(C)C(CC=C)C(=O)NC(=O)N |
C9H16N2O2 | |
MFCD00210239 | |
Apronalide, Allylisopropylacetylcarbamide, Allylisoprorylacetylurea, (2-Isopropyl-4-pentenoyl)urea, N-Carbamoyl-2-isopropyl-4-pentenamide | |
N-carbamoyl-2-propan-2-ylpent-4-enamide |
Specifications
528-92-7 | |
White | |
MFCD00210239 | |
Apronalide, Allylisopropylacetylcarbamide, Allylisoprorylacetylurea, (2-Isopropyl-4-pentenoyl)urea, N-Carbamoyl-2-isopropyl-4-pentenamide | |
CC(C)C(CC=C)C(=O)NC(=O)N | |
184.239 | |
184.24 | |
Crystalline Powder |
196°C | |
C9H16N2O2 | |
5 g | |
KSUUMAWCGDNLFK-UHFFFAOYSA-N | |
N-carbamoyl-2-propan-2-ylpent-4-enamide | |
10715 | |
≥98.0% (HPLC,N) | |
Apronal |
Safety and Handling
EINECSNumber : (9)-0107
RTECSNumber : YT5775000
TSCA : No