Aqualine™ Water Standard 5.0, Karl Fischer, Fisher Chemical
5mg/mL H2O standard
Supplier:  Thermo Fisher Scientific AL2730500
Catalog No. AL-2730500
Explore available promotions
Description
Description
  • Standard contains 5mg H2O/mL
  • Used for titer determination of volumetric Karl Fischer reagents
Specifications
Specifications
500 mL
Karl Fischer Reagent
71-36-3,1330-20-7
Liquid
Aqualine™ Standard 5.1
4.8 to 5.2mg of water/mL
Amber Glass
MFCD00077264