missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Aquasearcher™ pH Meter AB23PH

The Aquasearcher™ pH Meter AB23PH is a simple to use benchtop meter for measuring pH and oxidation-reduction potential (ORP). An intuitive design and auto-buffer recognition make measuring pH straightforward and easy.

Supplier:  OHAUS™ 30589821

 View more versions of this product

Catalog No. 01-922-374


Explore available promotions

Description

Description

  • Equipped with a large 5 in. LCD display and six instructional menu buttons
  • Auto buffer recognition and 3-point calibration capability ensures a reliable calibration result
  • Features a 99 item memory for measurements and last calibration
  • Choose between just the AB23PH meter with electrode holder and a full kit including AB23PH meter, ST320 pH electrode, and four pH Buffer Mini Kits (50 mL)
Specifications

Specifications

±0.01
±1
AB23PH Meter, Electrode Holder, ST320 pH Electrode, 4 pH Buffer Mini Kits (50 mL)
Linear
0.01
Up to 3
LCD, 5 in. Segment Matrix
99 Sets
Datalog for Measurements and Calibration
±0.5°C
Auto-stop, Continuous
0 to 14
±1999
0.0°C to 100.0°C
LCD
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions