missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
OHAUS™ Aquasearcher™ pH Meter AB41PH
The Aquasearcher™ pH Meter AB41PH is an advanced, research-grade benchtop pH meter with electrode, offering accurate, repeatable results with selectable resolution and an intelligent i-Steward system.
Supplier: OHAUS™ 30589831
Description
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory and password management for GLP documentation
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward ensures repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)
Specifications
±0.002 | |
USB, RS-232 (PC), RS-232 (Printer) | |
±0.3°C | |
Auto-stop, Timed, Continuous | |
-2 to 20 | |
±2000.0 | |
-10°C to +125°C | |
LCD | |
1000 Measurement, 10 Calibration |
Datalog with Time and Date Stamp | |
±0.3 | |
AB41PH Meter, ST410 Electrode, STTEMP Probe, Electrode Holder, AS20 Magnetic Stirrer, 4 pH Buffer Mini Kits (50 mL) | |
Offset | |
0.001, 0.01, 0.1 | |
Up to 5 | |
LCD, 6.5 in. Segment and Dot Matrix | |
1000 Sets |