missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wells Lamont™ Aramid Fiber Sleeves
These 2 ply Aramid Fiber Sleeves are an ideal solution in providing arm protection and comfort. ANSI A3 rating provides great cut protection, and the material made from para-aramid yarns adds safety with flame-resistant properties. Offer excellent protection against cutting and burn hazards.
Supplier: Wells Lamont™ SK14
Description
- 2-ply knit sleeve
- Flame resistant
- Available with or without a thumb hole
- ANSI Level 2 Heat Rating
- ANSI Level 4 Flame Rating
- ANSI Cut Level A3
- Made in the USA
Specifications
2-ply | |
Aramid Fiber | |
Standard Sleeve | |
Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution. | |
35.5 cm |
Knit Sleeve | |
Yellow | |
Reusable | |
14 in. |