Wells Lamont™ Aramid Fiber Sleeves
These 2 ply Aramid Fiber Sleeves are an ideal solution in providing arm protection and comfort. ANSI A3 rating provides great cut protection, and the material made from para-aramid yarns adds safety with flame-resistant properties. Offer excellent protection against cutting and burn hazards.
Supplier:  Wells Lamont™ SK18T
 View more versions of this product
Catalog No. 19053941
Explore available promotions
Description
Description
  • 2-ply knit sleeve
  • Flame resistant
  • Available with or without a thumb hole
  • ANSI Level 2 Heat Rating
  • ANSI Level 4 Flame Rating
  • ANSI Cut Level A3
  • Made in the USA
Specifications
Specifications
Aramid Fiber with one track
Yellow
Reusable
Knit Sleeve
Sleeve with Thumb Hole
Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution.