National Safety Apparel™ Arc H2O FR Hi-Vis Yellow Rain Jacket - Type R Class 3
Arc H2O FR HI-Vis rain jacket type R class 3
Supplier: National Safety Apparel™ R30RL06SM
Description
- Arc Rating: 19 cal/cm² (Fluorescent Yellow) | 24 cal/cm² (Fluorescent Orange)
- CAT 2
- Fabric/Material: PU Coated FR Cotton
- Waterproof welded seams with reinforced stress points to provide durability
- FR fabric will not melt, drip or ignite
- FR polyurethane is durable and flexible that will not become brittle as it ages
- Contoured collar with tuck away attached hood
- Double storm flap with reflective trim
- Adjustable cuffs
- Back ventilation system with FR mesh for increased airflow
- Covered D-ring opening for opening access
- Two reinforced front pockets with closure for convenient secure storage
- 2 in. silver reflective trim
- FR hook and loop tab at wrist cuff for improved fit
- Adjustable elastic suspender system for fit and comfort
- Covered zipper and snap closures
- Made in the USA
Specifications
Small | |
Fluorescent Yellow | |
Arc Rating: 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange), CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269 |
Polyurethane-coated FR Cotton | |
Snap, Zipper | |
Welded |