missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
National Safety Apparel™ ArcGuard™ Aramid Fiber Arc Flash Gloves
Constructed from multiple layers of DuPont™ Nomex and aramid fiber that is inherently flame resistant and will not melt, drip, or ignite.
$381.79 - $567.66
Specifications
Certifications/Compliance | NFPA 70E, ASTM F1506, OSHA 1910.269 |
---|---|
Applications | Industrial |
Color | Brown |
Material | Nomex, Aramid Fiber |
Product Type | Arc Flash Gloves |
Description
- Constructed from multiple layers of DuPont™ Nomex™ and Kevlar™ Aramid Fiber
- Materials are inherently flame resistant and will not melt, drip, or ignite
- Lined with needle-punched DuPont™ Nomex™ felt for additional flame resistance
- Wide bell cuff for easy donning and doffing
Specifications
NFPA 70E, ASTM F1506, OSHA 1910.269 | |
Brown | |
Arc Flash Gloves | |
Arc Flash Hazard Protection | |
G51KDQ | |
45.7 cm |
Industrial | |
Nomex, Aramid Fiber | |
Universal | |
Open | |
18 in. |