missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
National Safety Apparel™ ArcGuard™ Performance Arc Flash Lift Front Hood, 40 Cal
ArcGuard™ Performance Arc Flash Lift Front Hood, 40 Cal, CAT4, PureView™ face shield, Anti-Fog and Scratch Resistant, One Size
Supplier: National Safety Apparel™ ARC40H
Description
- Single-layer 40 cal arc flash protection
- Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
- Using GORE™ PYRAD™ comfort science to keep the wearer cool, dry, comfortable, and unrestricted while working
- PureView™ face shield technology features a greater range of visibility
- Lift front shield makes it easy to obtain fresh air while out of the hazard zone
- Ergonomic design for ease of movement and flexibility
- Compliant to NFPA 70E for Arc Flash
- Compliant to CSA Z462 for Arc Flash (Canada)
- Compliant with OSHA 1910.269
Specifications
Arc Flash Hood | |
GORE Pyrad | |
Pull-over | |
CAT 4, NFPA 70E, CSA Z462, OSHA 1910.269 | |
Hard hat, face shield, hood | |
Arc Flash PPE | |
Lift Front Hood |
Universal | |
Navy | |
Reusable | |
9.5 oz. | |
Hood | |
Lift Front |