missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arecoline hydrobromide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 250130250

 View more versions of this product

Catalog No. AC250130250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Arecoline hydrobromide
171°C to 174°C
Authentic
Glass bottle
MFCD00039041
15, 768
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in chloroform and ether
[H+].[Br-].COC(=O)C1=CCCN(C)C1
236.11
236.11
Powder
300-08-3
White to Yellow
98% min. (Argentometry)
C8H14BrNO2
25 g
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
9301
98+%

Chemical Identifiers

300-08-3
236.11
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
9301
[H+].[Br-].COC(=O)C1=CCCN(C)C1
C8H14BrNO2
MFCD00039041
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-087-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only