Learn More
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis), Thermo Scientific Chemicals
$47.30 - $8016.92
Chemical Identifiers
CAS | 7440-38-2 |
---|---|
Molecular Formula | As |
Molecular Weight (g/mol) | 74.92 |
MDL Number | MFCD00085309 |
InChI Key | RQNWIZPPADIBDY-UHFFFAOYSA-N |
Synonym | arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal |
PubChem CID | 5359596 |
ChEBI | CHEBI:27563 |
IUPAC Name | arsenic |
SMILES | [As] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1304403
|
Thermo Scientific Chemicals
01304403 |
1 g |
Each for $47.30
|
|
|||||
AA1304409
|
Thermo Scientific Chemicals
01304409 |
10 g |
Each for $273.93
|
|
|||||
AA1304414
|
Thermo Scientific Chemicals
01304414 |
25 g |
Each for $436.45
|
|
|||||
AA1304422
|
Thermo Scientific Chemicals
01304422 |
100 g |
Each for $1,073.79
|
|
|||||
AA13044A1
|
Thermo Scientific Chemicals
013044A1 |
1 kg |
Each for $8,016.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-38-2 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
5359596 | |
arsenic |
As | |
MFCD00085309 | |
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal | |
CHEBI:27563 | |
[As] |
Specifications
Gray | |
As | |
UN1558 | |
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal | |
[As] | |
74.92 | |
CHEBI:27563 | |
≥99.99999% | |
Odorless | |
Arsenic polycrystalline lump, 2 to 8mm (0.08 to 0.3 in.) |
7440-38-2 | |
MFCD00085309 | |
1 g | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
74.92 | |
Puratronic™ | |
Air Sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC
EINECSNumber : 213-148-6
RTECSNumber : CG0525000
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only