Learn More
Thermo Scientific™ ART™ Wide Bore Filtered Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
$470.17 - $566.57
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Genomic/Wide Orifice |
Catalog Number | Mfr. No. | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price | Quantity | ||
21-236-1A
|
Thermo Scientific™
2069G |
ART 200G, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Genomic/Wide Orifice | Sterile & PCR Ready |
Pack of 960 Each for $566.57
Case of 5 Pack for $2,832.85
|
|
||
21-236-2C
|
Thermo Scientific™
2079G |
ART 1000G, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Genomic/Wide Orifice | Sterile & PCR Ready |
Pack of 800 Each for $470.17
Case of 4 Pack for $1,880.68
|
|
||
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
- Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
- Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Filtered | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Genomic/Wide Orifice | |
ART |
Sterile | |
Lift-off Lid Rack | |
Universal Fit | |
Sterile & PCR Ready |