MilliporeSigma™ Supelco™ Ascentis™ Express 90 Å PFAS, 2.7 μm HPLC Column
The densely bonded, extensively endcapped ODS stationary phase of Ascentis™ Express PFAS provides an application tested solution for PFAS analysis.
Supplier:  MilliporeSigma™ Supelco™ 53560U
 View more versions of this product
Catalog No. 111023079
Explore available promotions
Description
Description
  • Ascentis™ Express PFAS is a high-speed, high-performance liquid chromatography column based on the proven Fused-Core™ particle design.
  • The Fused-Core™ particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
  • This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thick porous shell and the small overall particle size of 2.7 μm.
  • The densely bonded, extensively endcapped ODS stationary phase of Ascentis™ Express PFAS provides an application tested solution for PFAS analysis.
Specifications
Specifications
2.1 mm
600 bar
2 to 9
1 Ea.
<60°C
Reversed Phase
HPLC, LC/MS, UHPLC, UHPLC-MS
15 cm
2.7 μm
90 Å
135 m2/g
Fused-Core™ Silica Particle Platform Particle Platform Superficially Porous Particle
Stainless Steel