missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aspartame, Powder, NF, 98-102%, Spectrum™ Chemical
A1378, 22839-47-0, C14H18N2O5
$224.90 - $7864.50
Chemical Identifiers
CAS | 22839-47-0 |
---|---|
Molecular Formula | C14H18N2O5 |
Molecular Weight (g/mol) | 294.31 |
MDL Number | MFCD00002724 |
InChI Key | IAOZJIPTCAWIRG-QWRGUYRKSA-N |
IUPAC Name | (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid |
SMILES | COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18600797
|
Spectrum Chemical Mfg Cor
A137825GM |
25 g | Poly Bottle |
Each for $224.90
|
|
||||
18600798
|
Spectrum Chemical Mfg Cor
A1378125GM |
125 g | Poly Bottle |
Each for $594.48
|
|
||||
18600799
|
Spectrum Chemical Mfg Cor
A1378500GM |
500 g | Poly Pail |
Each for $1,613.35
|
|
||||
18600801
|
Spectrum Chemical Mfg Cor
A137812KGBL |
12 kg | Fiber Drum |
Each for $6,218.04
|
|
||||
18600800
|
Spectrum Chemical Mfg Cor
A137825KGBL |
25 kg | Fiber Drum |
Each for $7,864.50
|
|
||||
Description
Spectrum™ Chemical Aspartame, Powder, NF is used in pharmaceutical products, often as a sugar replacement in chewable tablets and sugar-free liquids. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
22839-47-0 | |
294.31 | |
IAOZJIPTCAWIRG-QWRGUYRKSA-N | |
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O |
C14H18N2O5 | |
MFCD00002724 | |
(3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid |
Specifications
22839-47-0 | |
0.2% | |
MFCD00002724 | |
Poly Bottle | |
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O | |
+14.5° to +16.5° | |
98 to 102% | |
25 g |
100% | |
C14H18N2O5 | |
4.5% | |
IAOZJIPTCAWIRG-QWRGUYRKSA-N | |
(3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid | |
294.31 | |
NF |