missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medicom™ AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips
AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips. Ideal for procedures that require a high level of precision and tactile sensitivity. DermaSense technology helps reduce the risk of skin reactions. Textured fingertips ensure a secure grip. Chemo-tested.
Supplier: Medicom™ 9970A
Description
- These chemo-tested, ultra-thin nitrile gloves are ideal for procedures that require precision and tactile sensitivity.
- Textured fingertips ensure a secure grip, even when wet.
- DermaSense technology eliminates the chemical accelerators commonly found in nitrile gloves to reduce the risk of skin reactions.
Specifications
No | |
Healthcare, Industrial, Laboratory | |
Blue | |
Fingertip Textured | |
Powder Free | |
Non-sterile | |
Beaded |
Yes | |
1.5 | |
Single-use | |
Yes | |
X-Small | |
Yes |