missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medicom™ AssureWear™ White SMS Lab Coats
Non-woven SMS lab coat with knitted cuffs is designed to protect healthcare professionals, lab technicians, and other wearers from fluids and cross-contamination.
Supplier: Medicom™ A8042
Description
- Non-woven 3-layer SMS
- Knit cuffs
- Knit collar
Specifications
General Purpose Lab Coat | |
Unisex | |
White | |
Knit | |
Non-sterile | |
None | |
Yes | |
Healthcare, Laboratories |
Medium | |
SMS | |
Snaps | |
Single-use | |
Knee-length | |
Knit | |
A8042 | |
Not Autoclavable |