Troemner™ ASTM Class 2 Weight with ISO/IEC 17025 Accredited Certificate
Troemener Individual Calibration Weights are Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
Supplier:  Troemner™ 80858259
 View more versions of this product
Catalog No. 02218216
Explore available promotions
Description
Description
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
Specifications
Specifications
Calibration Mass
Troemner Alloy 8
5 g
ISO/IEC 17025 Accredited Certificate
Polycarbonate
Cylindrical