missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Atenolol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 449550010

 View more versions of this product

Catalog No. AC449550010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Atenolol
146.0°C to 148.0°C
Glass bottle
1 g
atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
METKIMKYRPQLGS-UHFFFAOYSA-N
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
2249
266.34
29122-68-7
98%
C14H22N2O3
15,85
Solubility in water: slightly soluble. Other solubilities: soluble in methanol, acetic acid, dmso, ethanol, insoluble in acetonitrile, ethylacetate, chlorofor, m
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
266.34
CHEBI:2904
98%

Chemical Identifiers

29122-68-7
266.34
atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
CHEBI:2904
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
C14H22N2O3
METKIMKYRPQLGS-UHFFFAOYSA-N
2249
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 249-451-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only