missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Atenolol 98.0+%, TCI America™

Supplier:  TCI America A141410G

Catalog No. A141410G


Explore available promotions

Description

Description

Specifications

Specifications

Atenolol
154°C
C14H22N2O3
10 g
METKIMKYRPQLGS-UHFFFAOYSA-N
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
2249
266.34
Crystalline Powder
29122-68-7
White-Yellow
MFCD00057645
atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
266.341
CHEBI:2904
≥98.0% (T)

Chemical Identifiers

29122-68-7
266.341
METKIMKYRPQLGS-UHFFFAOYSA-N
2249
2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
C14H22N2O3
MFCD00057645
atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
CHEBI:2904
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
Videos
Safety and Handling

Safety and Handling

RTECSNumber : AC3600000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions