3M™ N95 Aura™ Health Care Particulate Respirator and Surgical Mask, 1870+
Comfortable inner materials provide respiratory protection against certain airborne particles. Individually packaged flat-fold design allows for convenient storage.
Supplier:  3M™ 7100235156
Catalog No. 296368419
Explore available promotions
Alert:
WARNING: These respirators help reduce exposures to certain airborne particulates. Before use, wearer must read and understand the user instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting the requirements of the jurisdiction and/or CSA Z94.4 respiratory standard. Misuse may result in sickness or death. For proper use, see package instructions, supervisor, or call 3M™ PSD Technical Service in Canada at 1-800-267-4414 and in USA at 1-800-243-4630.
Description
Description
  • Healthcare respirator meets CDC guidelines for M. tuberculosis exposure control
  • Disposable particulate respirator intended to reduce exposure to airborne particles including those generated by electrocautery, laser surgery and other powered medical instruments
  • Surgical mask designed to be fluid-resistant to splash and spatter of blood and other infectious materials
Note:
  • Before use, wearer must read and understand user instructions provided as part of the product packaging
Specifications
Specifications
N - Not Resistant to Oil
Health Canada Class I Medical Device
OSHA 1910.134
Yes
Polyurethane, Polypropylene
Standard
95%
Polyisoprene
Aluminum
Individually Packaged
Fluid 160 mmHg
No
Yes
10
NIOSH 42 CFR 84, N95, approval number: TC-84A-5726
Flat-fold
N95
Healthcare Particulate Respirator and Surgical Mask
No
Yes
1870+
Adjustable Nose Clip
440/Cs.
Non-sterile
Stapled Headbands
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'