missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aushon Chemiluminescent Human Cytokine Array Kits
For simultaneous measurement of combinations of cytokines
$1232.51 - $2375.63
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of combinations of cytokines |
Product Type | Cytokine Array Kits |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
020071FA
|
Aushon Biosystems Inc
020071FA |
3 Plex |
Each for $1,232.51
|
|
|||||
02FF81FA
|
Aushon Biosystems Inc
02FF81FA |
9 Plex |
Each for $2,375.63
|
|
|||||
021981FA
|
Aushon Biosystems Inc
021981FA |
4 Plex |
Each for $1,232.51
|
|
|||||
027681FA
|
Aushon Biosystems Inc
027681FA |
6 Plex |
Each for $1,801.37
|
|
|||||
028981FA
|
Aushon Biosystems Inc
028981FA |
4 Plex |
Each for $1,422.13
|
|
|||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating cytokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of cytokines | |
Cytokine Array Kits |