Aushon Chemiluminescent Human Growth Factor 3 Array Kits
For simultaneous measurement of combinations of growth factors
Supplier: Aushon Biosystems Inc 121041FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of growth factors | |
2 Plex |