Aushon Chemiluminescent Mouse Chemokine Array Kits DFS Item
For simultaneous measurement of combinations of chemokines
$876.65 - $2629.96
Specifications
For Use With (Application) For simultaneous measurement of combinations of chemokines
Product Type Chemokine Array Kits
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
190011FA DFS Item
View Documents
Aushon Biosystems Inc
190011FA
1 Plex
Each for $876.65
195981FA DFS Item
View Documents
Aushon Biosystems Inc
195981FA
5 Plex
Each for $1,655.90
19A641FA DFS Item
View Documents
Aushon Biosystems Inc
19A641FA
5 Plex
Each for $1,655.90
19FFC1FA DFS Item
View Documents
Aushon Biosystems Inc
19FFC1FA
10 Plex
Each for $2,629.96
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of chemokines
Chemokine Array Kits
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'