Aushon Chemiluminescent Mouse Cytokine Array Kits DFS Item
For simultaneous measurement of combinations of cytokines
Supplier:  Aushon Biosystems Inc 178851FA
 View more versions of this product
Catalog No. 178851FA
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of cytokines
Cytokine Array Kits
4 Plex