Aushon Chemiluminescent Mouse Cytokine Array Kits
For simultaneous measurement of combinations of cytokines
Supplier: Aushon Biosystems Inc 17FF51FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For simultaneous measurement of combinations of cytokines | |
Cytokine Array Kits | |
10 Plex |