Aushon Chemiluminescent Rat Chemokine Array Kits DFS Item
For simultaneous measurement of combinations of chemokines
$1461.10 - $2629.96
Specifications
For Use With (Application) For simultaneous measurement of combinations of chemokines
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
242581FA DFS Item
View Documents
Aushon Biosystems Inc
242581FA
4 Plex
Each for $1,461.10
243FF1FA DFS Item
View Documents
Aushon Biosystems Inc
243FF1FA
10 Plex
Each for $2,629.96
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of chemokines
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'