Axygen™ 1000μL Universal Pipetter Tips: Wide Bore, Maxymum Recovery
Compatible with most popular brands of pipettes
Supplier:  Axygen™ T1005WBCLR
Catalog No. 14-222-700
Explore available promotions
Description
Description
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Orifice size of 0.065 in. (1.6mm) makes wide bore tips suitable for pipetting viscous materials, genomic DNA or delicate cell culturese
  • Volume range: 100 to 1000μL
  • Available in racked format
Specifications
Specifications
100 to 1000 μL
Clear
DNase, RNase, DNA, and PCR inhibitor Free
Low Retention, Wide Bore
Polypropylene
Non-sterile
Autoclavable
Rack
Universal Fit