missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ 200μL Universal Pipette Tips, Beveled, Reference, Maxymum Recovery
Designed to fit a wide variety of single and multi-channel pipettes
$102.78 - $1003.83
Specifications
Volume (Metric) | 1 to 200 μL |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Tip Style | Beveled, Low Retention, Universal |
For Use With (Equipment) | Universal Fit |
Catalog Number | Mfr. No. | Format | Sterility | Color | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Sterility | Color | Price | Quantity | |||
14-222-812
|
Axygen™
TR222CL |
Bulk | Non-sterile | Clear |
Pack of 1000 Each for $102.78
Case of 20 Pack for $2,055.60
|
|
|||
14-222-813
|
Axygen™
TR222CLR |
Rack | Non-sterile | Clear |
Case of 960 Each for $950.65
|
|
|||
14-222-814
|
Axygen™
TR222CLRS |
Rack | Sterile | Clear |
Pack of 960 Each for $202.91
Case of 5 Pack for $1,014.55
|
|
|||
14-223-386
|
Axygen™
TR222CLSTKS |
Rack | Sterile | Clear |
Case of 4800 Each for $1,003.83
|
|
|||
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Specifications
1 to 200 μL | |
Autoclavable | |
Universal Fit |
Clear | |
Beveled, Low Retention, Universal | |
Polypropylene |