missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
Designed to fit a wide variety of single and multi-channel pipettes.
Supplier: Axygen™ T200CSTK
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
- Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents
Specifications
200 μL | |
Non-sterile | |
Not Autoclavable | |
Rack, 5 x 96 | |
Polypropylene |
Non-Filtered | |
Clear | |
DNase, RNase Free | |
Universal Fit, Universal Fit |