missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ 96-well PCR Microplates

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

$149.31 - $945.00

Specifications

Barcode No Barcode
Certifications/Compliance RNase-, DNase-free
Format Standard
Array 8 x 12
Material Polypropylene
View More Specs

Products 8
Catalog Number Mfr. No. Color Skirt Style For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color Skirt Style For Use With (Equipment) Price Quantity  
14-222-320
View Documents Promotion Details
Axygen™
PCR96ABC
Clear Elevated ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
Pack of 10 Each for $156.14
Case of 5 Pack for $780.70
 
14-222-321
View Documents Promotion Details
Axygen™
PCR96C
Clear Elevated Statagene MX4000
Pack of 10 Each for $151.38
Case of 5 Pack for $756.90
 
14-222-322
View Documents Promotion Details
Axygen™
PCR96FLTC
Clear Unskirted BioRad qPCR systems, Statagene MX-3000
Pack of 25 Each for $232.24
Case of 4 Pack for $928.96
 
14-222-326
View Documents Promotion Details
Axygen™
PCR96FSC
Clear Full Standard 96 well blocks
Pack of 10 Each for $162.24
Case of 5 Pack for $811.20
 
50-400-611
View Documents Promotion Details
Axygen™
PCR96FSB
Blue Full Standard 96 well blocks
Case of 50 Each for $945.00
 
50-400-641
View Documents Promotion Details
Axygen™
PCR96MBC
Clear Unskirted Megabace Sequencer
Case of 50 Each for $814.07
 
14-222-334
View Documents Promotion Details
Axygen™
PCR96HSC
Clear Half Standard 96 well blocks
Pack of 10 Each for $212.83
Case of 5 Pack for $1,064.15
 
14-222-340
View Documents Promotion Details
Axygen™
PCR96M2HSC
Clear Half ABI PCR 2700, 2720, 9700, 9800, 7300, 7500, 7900 and ABI sequencing units
Pack of 10 Each for $149.31
Case of 5 Pack for $746.55
 
Description

Description

Axygen™ 96 well PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • Well Volume: 200 μL
Specifications

Specifications

No Barcode
Standard
Polypropylene
96
PCR thermal transfer, RT-PCR applications
RNase-, DNase-free
8 x 12
200 μL
Non-sterile
SDS
missing translation for 'documents'

missing translation for 'documents'