missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Gel Casting Trays for 7 cm Gel Box

Accessory for use with 7cm horizontal gel box

Supplier:  Axygen™ HGB77GT

 View more versions of this product

Catalog No. 14223963


Explore available promotions

Description

Description

  • UV transparent
Specifications

Specifications

Gel Box, 7 cm
7 x 7 cm (2.7 x 2.7 in.)
Videos
SDS
Documents

Documents

Promotions

Promotions