missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ Multi Well Low Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
$771.16 - $1023.75
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Material | Polypropylene |
Type | Reagent Reservoir |
No. per Case | 25/Cs. |
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity | ||
50-400-718
|
Axygen™
RESMW12LP |
84 mL | Non-sterile | 12 | 25/Cs. |
Case of 25 Each for $771.16
|
|
||
14-222-405
|
Axygen™
RESMW12LPSI |
21 mL | Sterile | 12 | Individually Wrapped; 25/Cs. |
Case of 25 Each for $880.38
|
|
||
14-222-409
|
Axygen™
RESMW8LP |
32 mL | Non-sterile | 8 | 25/Cs. |
Case of 25 Each for $790.84
|
|
||
14-222-410
|
Axygen™
RESMW8LPSI |
32 mL | Sterile | 8 | Individually Wrapped; 25/Cs. |
Case of 25 Each for $1,023.75
|
|
||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
Polypropylene | |
25/Cs. |
Clear | |
Reagent Reservoir | |
Single-use |