Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Supplier: Axygen™ RFL222C
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.Specifications
1 to 200 μL | |
Clear | |
Universal | |
Yes |
Non-sterile | |
Autoclavable | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs |