Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Supplier: Axygen™ RFL300C
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.Specifications
0.1 to 10 μL | |
Clear | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
No |
Non-sterile | |
Autoclavable | |
Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2, Universal Fit |