Azer Scientific Choyce Clean Catch Urine Collection Kit
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics
$332.71 - $605.44
Specifications
For Use With (Application) | Used to conduct culture and sensitivity studies for determining efficacy of antibiotics |
---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
21-100-700
|
Azer Scientific
ES63321 |
Urine Collection Kit |
Case of 50 Each for $332.71
|
|
|||||
21-100-701
|
Azer Scientific
ES64310 |
Grip Handle |
Case of 500 Each for $605.44
|
|
|||||
Description
- Kit (21-100-700): contains one sterile 90mL/53mm urine collection cup and includes Azer Grip handle to prevent contamination, two castile soap towelettes and instructions in Spanish and English
- Grip Handles (21-100-701): can be purchased separately in cases of 500
Specifications
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics |