missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

Designed specifically for the collection and transport of various sizes of tissue samples

Supplier:  Azer Scientific BPFNBF90

 View more versions of this product

Catalog No. 22026442


Explore available promotions

Description

Description

  • A traditional fixative that is buffered to a neutral pH
  • Ideal for histological applications for fixing and storing tissue specimens
  • Each container is half filled with 10 percent neutral buffered formalin
  • Packaged with our proprietary Sure-Lok closure

Compliance

Containers are 95 kPa compliant.

Specifications

Specifications

90 mL
12 Pk., 20 Pks/Cs.
Fixative, safe collection, preservation, transportation, and storage of specimens of various sizes
45mL Neutral Buffered Formalin 10%
Formalin
Container
Videos
SDS
Documents

Documents

Promotions

Promotions