missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azetidine, 98%, Thermo Scientific Chemicals

$227.50 - $606.27

Chemical Identifiers

CAS 503-29-7
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.09
InChI Key HONIICLYMWZJFZ-UHFFFAOYSA-N
Synonym azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
PubChem CID 10422
ChEBI CHEBI:30968
IUPAC Name azetidine
SMILES C1CNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296732500
SDS View Documents
Thermo Scientific Chemicals
296732500
250 mg Glass bottle
Each for $227.50
 
AC296730010
SDS View Documents
Thermo Scientific Chemicals
296730010
1 g Glass bottle
Each for $606.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

503-29-7
57.09
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
CHEBI:30968
C1CNC1
C3H7N
HONIICLYMWZJFZ-UHFFFAOYSA-N
10422
azetidine

Specifications

503-29-7
0.8500g/mL
−20°C
97.5% min. (GC)
C3H7N
250 mg
0.85
Solubility in water: miscible.
C1CNC1
57.09
CHEBI:30968
98%
Azetidine
Colorless
61.0°C to 62.0°C
Authentic
Glass bottle
1.4290 to 1.4330
20,2
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
HONIICLYMWZJFZ-UHFFFAOYSA-N
azetidine
10422
57.09
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 207-963-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only