missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-biotin conjugate

An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media | CAS: 875770-34-6 | C18H32N6O5S | 444.551 g/mol

$312.47 - $633.74

Chemical Identifiers

CAS 875770-34-6
Molecular Formula C18H32N6O5S
Molecular Weight (g/mol) 444.551
MDL Number MFCD20134145
InChI Key ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
Synonym biotin-peg3-azide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl biotinamide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamide, biotin-peg4-n3, azide-peg3-biotin conjugate, azido-peg3-biotin conjugate, 11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane, 3abeta,6abeta-n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-2-oxohexahydro-1h-thieno 3,4-d imidazole-4alpha-pentanamide
PubChem CID 60146223
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64996MB
View Documents
Thermo Scientific Chemicals
J64996MB
25 mg
Each for $312.47
Only null left
 
AAJ64996MC
View Documents
Thermo Scientific Chemicals
J64996MC
100 mg
Each for $633.74
Only null left
 
Description

Description

Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it?s components do not react with the biological environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Solubility
Solubile in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

875770-34-6
444.551
ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
60146223
C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2
C18H32N6O5S
MFCD20134145
biotin-peg3-azide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl biotinamide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamide, biotin-peg4-n3, azide-peg3-biotin conjugate, azido-peg3-biotin conjugate, 11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane, 3abeta,6abeta-n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-2-oxohexahydro-1h-thieno 3,4-d imidazole-4alpha-pentanamide
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide

Specifications

875770-34-6
C18H32N6O5S
25 mg
Solubile in DMSO,DMF and water.
C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2
444.551
444.55
Azido-PEG3-biotin conjugate
White to Gray
MFCD20134145
biotin-peg3-azide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl biotinamide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamide, biotin-peg4-n3, azide-peg3-biotin conjugate, azido-peg3-biotin conjugate, 11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane, 3abeta,6abeta-n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-2-oxohexahydro-1h-thieno 3,4-d imidazole-4alpha-pentanamide
ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
60146223
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only