missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azido-PEG4-Amine 95.0+%, TCI America™

$128.38 - $387.10

Chemical Identifiers

CAS 951671-92-4
Molecular Formula C10H22N4O4
Molecular Weight (g/mol) 262.31
MDL Number MFCD20134134
InChI Key ZMBGKXBIVYXREN-UHFFFAOYSA-N
Synonym Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane
PubChem CID 71313220
IUPAC Name 14-azido-3,6,9,12-tetraoxatetradecan-1-amine
SMILES NCCOCCOCCOCCOCCN=[N+]=[N-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A300450MG
View Documents
TCI America
A300450MG
50 mg
Each for $128.38
 
A3004250MG
View Documents
TCI America
A3004250MG
250 mg
Each for $387.10
 
Description

Description

Specifications

Chemical Identifiers

951671-92-4
262.31
ZMBGKXBIVYXREN-UHFFFAOYSA-N
71313220
NCCOCCOCCOCCOCCN=[N+]=[N-]
C10H22N4O4
MFCD20134134
Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane
14-azido-3,6,9,12-tetraoxatetradecan-1-amine

Specifications

951671-92-4
C10H22N4O4
50 mg
Amino-PEG4-Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane
NCCOCCOCCOCCOCCN=[N+]=[N-]
262.31
262.31
Liquid
Yellow
MFCD20134134
2735
ZMBGKXBIVYXREN-UHFFFAOYSA-N
14-azido-3,6,9,12-tetraoxatetradecan-1-amine
71313220
≥95.0% (GC)
Azido-PEG4-Amine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
missing translation for 'documents'

missing translation for 'documents'