missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dynalon™ Azlon™ Polypropylene Carboy with Spigot
For Use With: Storing and dispensing liquids in educational settings.; Autoclavable; Closure Material: Polypropylene; Color: White
$214.56 - $359.77
Specifications
Autoclavable | Autoclavable |
---|---|
Closure Material | Polypropylene |
Color | White |
For Use With (Application) | Storing and dispensing liquids in educational settings. |
Includes | Blow molded handles |
Description
- Autoclavable polypropylene™, heavy-duty construction
- Stylish rounded octagonal shape for increased rigidity when full
- Offset neck for easier filling and pouring
- Integral, comfortable blow molded handles for ease of carrying, are end-sealed to prevent contamination
- Heavy-duty closures
- Carboys feature a durable and leakproof heavy-duty spigot with boss fitting for extra strength
- Spigot features removable tube fitting for added convenience
- Molded graduations allow easy viewing of contents
Specifications
Autoclavable | |
White | |
Blow molded handles | |
Offset | |
Rounded Octagonal |
Polypropylene | |
Storing and dispensing liquids in educational settings. | |
Polypropylene | |
Carboy with Spigot |