missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azobenzene, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1042514

 View more versions of this product

Catalog No. AAA1042514


Explore available promotions

Description

Description

Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
Specifications

Specifications

Azobenzene
65°C to 69°C
293°C
MFCD00003022
UN3077
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
diphenyldiazene
2272
182.23
103-33-3
1.203
C12H10N2
25 g
14,917
Soluble in alcohol,ether,benzene and glacial acetic acid. Insoluble in water.
C1=CC=C(C=C1)N=NC1=CC=CC=C1
182.23
CHEBI:190358
≥97%

Chemical Identifiers

103-33-3
182.23
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
2272
diphenyldiazene
C12H10N2
MFCD00003022
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
CHEBI:190358
C1=CC=C(C=C1)N=NC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H373-H302-H332
May cause cancer.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H340-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 203-102-5

RTECSNumber : CN1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only