Learn More
Azobenzene, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1042536
Description
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.
Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.
Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
Specifications
Azobenzene | |
65°C to 69°C | |
293°C | |
MFCD00003022 | |
UN3077 | |
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide | |
DMLAVOWQYNRWNQ-UHFFFAOYSA-N | |
diphenyldiazene | |
2272 | |
182.23 |
103-33-3 | |
1.203 | |
C12H10N2 | |
500 g | |
14,917 | |
Soluble in alcohol,ether,benzene and glacial acetic acid. Insoluble in water. | |
C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
182.23 | |
CHEBI:190358 | |
≥97% |
Chemical Identifiers
103-33-3 | |
182.23 | |
DMLAVOWQYNRWNQ-UHFFFAOYSA-N | |
2272 | |
diphenyldiazene |
C12H10N2 | |
MFCD00003022 | |
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide | |
CHEBI:190358 | |
C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H350-H341-H373-H302-H332
May cause cancer.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H340-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 203-102-5
RTECSNumber : CN1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only