Learn More
Azobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401540500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Azobenzene | |
66.0°C to 69.0°C | |
293.0°C | |
Authentic | |
Glass bottle | |
C6H5N=NC6H5 | |
50 g | |
15, 909 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, glacial acetic acid,, acetone and benzene | |
C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
182.23 | |
CHEBI:190358 | |
98% |
103-33-3 | |
Orange | |
100°C | |
97.5% min. (GC) | |
C12H10N2 | |
MFCD00003022 | |
16, 8 | |
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide | |
DMLAVOWQYNRWNQ-UHFFFAOYSA-N | |
diphenyldiazene | |
2272 | |
182.23 | |
Crystalline Powder |
Chemical Identifiers
103-33-3 | |
182.23 | |
DMLAVOWQYNRWNQ-UHFFFAOYSA-N | |
2272 | |
diphenyldiazene |
C12H10N2 | |
MFCD00003022 | |
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide | |
CHEBI:190358 | |
C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-102-5
RUO – Research Use Only