missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 401542500

 View more versions of this product

Catalog No. AC401542500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Azobenzene
66.0°C to 69.0°C
100g/mL
100°C
97.5% min. (GC)
C12H10N2
MFCD00003022
16, 8
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
diphenyldiazene
2272
182.23
Crystalline Powder
103-33-3
Orange
293.0°C
Authentic
Glass bottle
C6H5N=NC6H5
250 g
15, 909
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, glacial acetic acid,, acetone and benzene
C1=CC=C(C=C1)N=NC1=CC=CC=C1
182.23
CHEBI:190358
98%

Chemical Identifiers

103-33-3
182.23
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
2272
diphenyldiazene
C12H10N2
MFCD00003022
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
CHEBI:190358
C1=CC=C(C=C1)N=NC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-102-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only