missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azomethine H, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215416910

 View more versions of this product

Catalog No. ICN15416910


Explore available promotions

Description

Description

Specifications

Specifications

Azomethine H
5941-07-1
Extinction Coefficient max: 237nm C=0.01% ≥50, 000 (Lit.)
MFCD00149588
BEDIAJJRAYEWBI-TTWKNDKESA-M
4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium
123134089
Orange
Powder
Chemistry
1.04
C17H14NNaO9S2
8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate
C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na]
463.41
423.4
10 g

Chemical Identifiers

5941-07-1
463.41
BEDIAJJRAYEWBI-TTWKNDKESA-M
C17H14NNaO9S2
MFCD00149588
8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-698-1

Recommended Storage : Store at 4°C.

SDS
Documents

Documents

Promotions

Promotions