missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aztreonam, Thermo Scientific Chemicals

$233.75 - $796.22

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
MDL Number MFCD00072145
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6288703
View Documents
Thermo Scientific Chemicals
J6288703
1 g
Each for $233.75
 
AAJ6288706
View Documents
Thermo Scientific Chemicals
J6288706
5 g
Each for $796.22
 
Description

Description

Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

Solubility
Soluble in DMF/water (1:1) at 50 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

78110-38-0
435.426
WZPBZJONDBGPKJ-PSGLRMFWSA-N
2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
C13H17N5O8S2
MFCD00072145
56842327
CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Aztreonam
78110-38-0
White
C13H17N5O8S2
14,925
WZPBZJONDBGPKJ-PSGLRMFWSA-N
2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
56842327
Aztreonam
Powder
227°C (decomposition)
1 g
MFCD00072145
Soluble in DMF/water (1:1) at 50 mg/ml
CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
435.426
435.43
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 278-839-9

RTECSNumber : UA2451400

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only