missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Azur eosin methylene-blue, pure, solution according to Giemsa
$42.76 - $108.27
Specifications
Chemical Name or Material | Azur eosin methylene-blue |
---|---|
CAS | 51811-82-6,67-56-1 |
MDL Number | MFCD00081642 |
Lambda Max | 523 to 526nm (c=0.1, MeOH) (eosine), 646 to 651nm (c=0.1, MeOH) (methylene blue) |
Grade | Pure |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC295591000
|
Thermo Scientific Chemicals
295591000 |
100 mL | Glass bottle |
Each for $42.76
|
|
||||
AC295595000
|
Thermo Scientific Chemicals
295595000 |
500 mL | Glass bottle |
Each for $108.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Azur eosin methylene-blue | |
MFCD00081642 | |
Pure | |
Blue |
51811-82-6,67-56-1 | |
523 to 526nm (c=0.1, MeOH) (eosine), 646 to 651nm (c=0.1, MeOH) (methylene blue) | |
Glass bottle | |
Solution |
RUO – Research Use Only